• USA Home
  • Search Results

  • scutellarein2862452953311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Scutellarein

2 Products
Synonym:   4′,5,6,7-Tetrahydroxyflavone, 6-Hydroxyapigenin
Image of Scutellarein
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O6
Molecular Weight:
 286.24
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More