• USA Home
  • Search Results

  • secbutylamine73141395284611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

sec-Butylamine

2 Products
Synonym:   (±)-2-Aminobutane, (±)-sec-Butylamine
Image of <I>sec</I>-Butylamine
CAS Number:
Linear Formula:
 CH3CH2CH(NH2)CH3
Molecular Weight:
 73.14
 Beilstein/REAXYS Number:
 1361345
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More