• USA Home
  • Search Results

  • secobarbital238287673311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Secobarbital

1 Products
Synonym:   5-(1-Methylbutyl)-5-(2-propenyl)-2,4,6(1H,3H,5H)-pyrimidinetrione, 5-Allyl-5-(1-methylbutyl)barbituric acid, Quinalbarbitone, Seconal
Image of Secobarbital
CAS Number:
Empirical Formula (Hill Notation):
 C12H18N2O3
Molecular Weight:
 238.28
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More