• USA Home
  • Search Results

  • selenium7896778249211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Selenium

12 Products
Image of Selenium
CAS Number:
Empirical Formula (Hill Notation):
 Se
Molecular Weight:
 78.96
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Weight
  Manufacturer designation
  Physical Form
  Particle Dimension
Show More

Product #
Description
229865 Selenium, powder, −100 mesh, 99.99% trace metals basis,
209651 Selenium, powder, −100 mesh, ≥99.5% trace metals basis,
209643 Selenium, pellets, <5 mm, ≥99.99% trace metals basis,
204307 Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis,
HT-SE101 Selenium, pellets, < 5mm, ≥99.999%,
GF66395212 Selenium, foil, 25x25mm, thickness 3mm, 99.95%,
GF10763456 Selenium, lump, 3 mm max. lump size, weight 20 g, high purity 99.999%,
GF33939849 Selenium, powder, max. particle size 250 micron, weight 200 g, purity 99.95%,
GF45431647 Selenium, lump, 3 mm max. lump size, weight 50 g, high purity 99.999%,
GF59784575 Selenium, powder, 250 max. part. size (micron), weight 50 g, purity 99.95%,
GF73071514 Selenium, powder, 250 max. part. size (micron), purity 99.95%, weight 100 g,
GF78955532 Selenium, lump, 3 mm max. lump size, weight 100 g, high purity 99.999%,