• USA Home
  • Search Results

  • sephadexg5012345900454011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sephadex® G-50

12 Products
CAS Number:
 MDL number:
 NACRES:
 NA.56
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Particle Dimension
  Physical Form
  pH Value
  Product Line
  Pore size
  Operating parameter
  Special Grade
  Feature
  Manufacturer designation
  Shelf Life
  Package quantity
  pH Range
  Swelling ratio
Show More