• USA Home
  • Search Results

  • sepharose6b12345901236611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sepharose® 6B

2 Products
Synonym:   Agarose, Beaded
Image of Sepharose<SUP>®</SUP> 6B
CAS Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Particle Dimension
  pH Range
  Pore size
Show More