• USA Home
  • Search Results

  • serotoninhydrochloride2126815398011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Serotonin hydrochloride

1 Products
Synonym:   3-(2-Aminoethyl)-5-hydroxyindole hydrochloride, 5-HT, 5-Hydroxytryptamine hydrochloride
Image of Serotonin hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C10H12N2O · HCl
Molecular Weight:
 212.68
 Beilstein/REAXYS Number:
 3570963
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More