• USA Home
  • Search Results

  • sertralinehydrochloride342697955997011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sertraline hydrochloride

4 Products
Synonym:   (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, Sertraline HCl
Image of Sertraline hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C17H17NCl2 · HCl
Molecular Weight:
 342.69
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
Show More