• USA Home
  • Search Results

  • sertralineimpuritye1521561171211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sertraline impurity E

1 Products
Synonym:   (R)-α-Hydroxyphenylacetic acid, (R)-(−)-Mandelic acid
Image of Sertraline impurity E
CAS Number:
Linear Formula:
 C6H5CH(OH)CO2H
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 2691094
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More