• USA Home
  • Search Results

  • sesamin3543560780711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sesamin

4 Products
Synonym:   Fagarol
Image of Sesamin
CAS Number:
Empirical Formula (Hill Notation):
 C20H18O6
Molecular Weight:
 354.35
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Physical Form
Show More