• USA Home
  • Search Results

  • sesamol1381253331311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sesamol

3 Products
Synonym:   3,4-(Methylenedioxy)phenol, 5-Benzodioxolol
Image of Sesamol
CAS Number:
Empirical Formula (Hill Notation):
 C7H6O3
Molecular Weight:
 138.12
 Beilstein/REAXYS Number:
 127405
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Product Line
Show More