• USA Home
  • Search Results

  • shikimicacid1741513859011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Shikimic acid

2 Products
Synonym:   (3R,4S,5R)-(−)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid
Image of Shikimic acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H10O5
Molecular Weight:
 174.15
 Beilstein/REAXYS Number:
 2210055
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Assay
Show More