• USA Home
  • Search Results

  • silicagel1234511292600811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silica gel

46 Products
Image of Silica gel
CAS Number:
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Particle Dimension
  Additive
  Chemical Composition
  Type
  Quality clarifier
  Assay
  Restriction
  Color
  ID clarifier
  Miscellaneous Specifications
  pH Value
  Manufacturer designation
  Surface Dimension
  Physical Form
  Pore size
  Special Grade
Show More

Product #
Description
236772 Silica gel, high-purity grade (Davisil Grade 633), pore size 60 Å, 200-425 mesh particle size,
60741 Silica gel, high-purity grade, average pore size 60 Å (52-73 Å), 70-230 mesh, 63-200 μm, for column chromatography,
60752 Silica gel, high-purity grade (w/ Ca, ~0.1%), pore size 60 Å, 230-400 mesh particle size,
236799 Silica gel, high-purity grade (Davisil Grade 635), pore size 60 Å, 60-100 mesh,
227196 Silica gel, high-purity grade (9385), pore size 60 Å, 230-400 mesh particle size,
236802 Silica gel, high-purity grade (Davisil Grade 636), pore size 60 Å, 35-60 mesh particle size,
214477 Silica gel, high-purity grade (Davisil Grade 923), pore size 30 Å, 100-200 mesh,
60737 Silica gel, high-purity grade, pore size 60 Å, 230-400 mesh particle size, 40-63 μm particle size, for flash chromatography,
391484 Silica gel, high-purity grade (7734), pore size 60 Å, 70-230 mesh,
S8394 Silica gel, packet of 5 g desiccant,
60738 Silica gel, high-purity grade, pore size 60 Å, 220-440 mesh particle size, 35-75 μm particle size, for flash chromatography,
288624 Silica gel, high-purity grade, pore size 60 Å, 70-230 mesh,
236756 Silica gel, Davisil® grade 710, pore size 50-76 Å, for thin layer chromatography,
346446 Silica gel, high-purity grade (7749), with gypsum binder and fluorescent indicator, for thin layer chromatography,
236810 Silica gel, high-purity grade (Davisil Grade 643), pore size 150 Å, 200-425 mesh,
645524 Silica gel, technical grade, pore size 60 Å, 200-425 mesh particle size,
717185 Silica gel, technical grade, pore size 60 Å, 230-400 mesh particle size, 40-63 μm particle size,
60765 Silica gel, high-purity grade, with ~15% calcium sulfate and fluorescent indicator, GF254, for thin layer chromatography,
S7500 Silica gel, Type II, 3.5 mm bead size, Suitable for desiccation,
717177 Silica gel, technical grade, pore size 60 Å, 70-230 mesh, 63-200 μm,
214396 Silica gel, high-purity grade (Davisil Grade 12), pore size 22 Å, 28-200 mesh,
236845 Silica gel, high-purity grade (Davisil Grade 646), 35-60 mesh, pore size 150 Å,
288594 Silica gel, high-purity grade, pore size 60 Å, 200-400 mesh particle size,
288608 Silica gel, high-purity grade, pore size 60 Å, 130-270 mesh, for column chromatography,
214426 Silica gel, technical grade 40, 6-14 mesh,
60735 Silica gel, high-purity grade, 40, 70-230 mesh, for column chromatography,
60775 Silica gel, high-purity grade, HF254, without calcium sulfate, with fluorescent indicator, for thin layer chromatography,
60740 Silica gel, high-purity grade (puriss), pore size 60 Å, 70-230 mesh, for column chromatography,
403598 Silica gel, high-purity grade (7754), pore size 60 Å, 70-230 mesh,
243981 Silica gel, high-purity grade (Davisil Grade 62), pore size 150 Å, 60-200 mesh,
60742 Silica gel, high-purity grade, 60Å, 35-60 mesh particle size,
12479 Silica gel, technical grade (w/ Ca, ~0.1%), 60Å, 230-400 mesh particle size, Ca 0.1-0.3 %,
343323 Silica gel, Davisil® grade 22, pore size 60 Å, 60-200 mesh,
288519 Silica gel, high-purity grade, pore size 60 Å, 5-25 μm particle size, without binder, for thin layer chromatography,
60736 Silica gel, high-purity grade, 40, 35-70 mesh, for column chromatography,
60745 Silica gel, high-purity grade, 90Å, 70-230 mesh, for column chromatography,
60760 Silica gel, high-purity grade, Type G, with ~13% calcium sulfate, for thin layer chromatography,
288586 Silica gel, high-purity grade, pore size 60 Å, 2-25 μm particle size, without binder, with fluorescent indicator, pore volume 0.75 cm3/g, for thin layer chromatography,
S6503 Silica gel, high-purity grade, Type G, 5-15 μm, for thin layer chromatography,
60770 Silica gel, high-purity grade, for thin layer chromatography, H, without calcium sulfate,
60750 Silica gel, high-purity grade, 40Å, 230-400 mesh, for preparative liquid chromatography,
60744 Silica gel, high-purity grade, 90Å, 15-25 μm, for column chromatography,
60746 Silica gel, high purity, 90Å, 35-70 mesh, for column chromatography,
288500 Silica gel, high-purity grade, pore size 60 Å, 2-25 μm particle size, without binder, pore volume 0.75 cm3/g, for thin layer chromatography,
03561 Silica gel, high-purity grade, FIA according to DIN 51791,
89943 Silica gel, for column chromatography, 60,