• USA Home
  • Search Results

  • silveracetate1669156363311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver acetate

4 Products
Synonym:   Acetic acid silver salt
Image of Silver acetate
CAS Number:
Linear Formula:
 CH3COOAg
Molecular Weight:
 166.91
 Beilstein/REAXYS Number:
 3595636
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More