• USA Home
  • Search Results

  • silvercarbonate2757553416711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver carbonate

2 Products
Image of Silver carbonate
CAS Number:
Linear Formula:
 Ag2CO3
Molecular Weight:
 275.75
 Beilstein/REAXYS Number:
 6936654
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  Physical Form
Show More