• USA Home
  • Search Results

  • silverchloride14332778390611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver chloride

3 Products
Synonym:   Chlorosilver
Image of Silver chloride
CAS Number:
Empirical Formula (Hill Notation):
 AgCl
Molecular Weight:
 143.32
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
Show More