• USA Home
  • Search Results

  • silverhexafluoroantimonatev343622604264811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver hexafluoroantimonate(V)

1 Products
Image of Silver hexafluoroantimonate(V)
CAS Number:
Linear Formula:
 AgSbF6
Molecular Weight:
 343.62
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More