• USA Home
  • Search Results

  • silveriodide23477778396211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver iodide

2 Products
Image of Silver iodide
CAS Number:
Empirical Formula (Hill Notation):
 AgI
Molecular Weight:
 234.77
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More