• USA Home
  • Search Results

  • silverioxide231742066712311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver(I) oxide

3 Products
Image of Silver(I) oxide
CAS Number:
Empirical Formula (Hill Notation):
 Ag2O
Molecular Weight:
 231.74
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  Restriction
  Special Grade
  Physical Form
Show More