• USA Home
  • Search Results

  • silverisulfadiazine357142219908211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver(I) sulfadiazine

1 Products
Synonym:   Silver sulfadiazine, Sulfadiazine silver
Image of Silver(I) sulfadiazine
CAS Number:
Empirical Formula (Hill Notation):
 C10H9AgN4O2S
Molecular Weight:
 357.14
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More