• USA Home
  • Search Results

  • silvernanowires10787744022411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver nanowires

3 Products
Synonym:   Silver nanofibers, Silver nanowhiskers, Silver nanowire
Image of Silver nanowires
CAS Number:
Empirical Formula (Hill Notation):
 Ag
Molecular Weight:
 107.87
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Density
  Concentration
  Composite Dimension
Show More