• USA Home
  • Search Results

  • silvernitrate16987776188811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver nitrate

13 Products
Synonym:   Nitric acid silver(I) salt
Image of Silver nitrate
CAS Number:
Linear Formula:
 AgNO3
Molecular Weight:
 169.87
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Suitability
  Miscellaneous Specifications
  Density
  Bulk Density
  Physical Form
  Product Line
  pH Value
  Purity Grade
  Solubility
  Special Grade
Show More

Product #
Description
209139 Silver nitrate, ACS reagent, ≥99.0%,
204390 Silver nitrate, 99.9999% trace metals basis,
S6506 Silver nitrate, ReagentPlus®, ≥99.0% (titration),
31630 Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%,
1.01512 Silver nitrate, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
S8157 Silver nitrate, BioXtra, >99% (titration),
10220 Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%,
1.01510 Silver nitrate, cryst. extra pure,
S7276 Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration),
85228 Silver nitrate, puriss. p.a., ≥99.5% (AT),
85229 Silver nitrate, tested according to Ph Eur,
SX0205 Silver nitrate, crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
792276 Silver nitrate, anhydrous, ≥99.999% trace metals basis,