• USA Home
  • Search Results

  • silversulfadiazine357142219908211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver sulfadiazine

2 Products
Synonym:   Silver(I) sulfadiazine, Sulfadiazine silver
Image of Silver sulfadiazine
CAS Number:
Empirical Formula (Hill Notation):
 C10H9AgN4O2S
Molecular Weight:
 357.14
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More