• USA Home
  • Search Results

  • silversulfate311801029426511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silver sulfate

8 Products
Synonym:   Sulfuric acid disilver(I) salt
Image of Silver sulfate
CAS Number:
Linear Formula:
 Ag2SO4
Molecular Weight:
 311.80
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
  Potency
  Density
  Bulk Density
  Restriction
  Solubility
  Purity Grade
  Melting Point
  Reaction Suitability
  pH Value
  Special Grade
  Physical Form
Show More