• USA Home
  • Search Results

  • silvex269519372111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silvex®

1 Products
Synonym:   2,4,5-TP, 2-(2,4,5-Trichlorophenoxy)propionic acid, Fenoprop, Silvex®
Image of Silvex<SUP>®</SUP>
CAS Number:
Empirical Formula (Hill Notation):
 C9H7Cl3O3
Molecular Weight:
 269.51
 Beilstein/REAXYS Number:
 1985768
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More