• USA Home
  • Search Results

  • silybinabmixture4824480291857611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silybin (A + B mixture)

1 Products
Synonym:   (2R,3R)-3,5,7-Trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-hydroxymethyl-2,3-dihydrobenzo[1,4]dioxin-6-yl]chroman-4-one, Silibinin, Silybin
Image of Silybin (A + B mixture)
CAS Number:
Empirical Formula (Hill Notation):
 C25H22O10
Molecular Weight:
 482.44
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More