• USA Home
  • Search Results

  • silydianin482442978268111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Silydianin

3 Products
Synonym:   Silidianin
Image of Silydianin
CAS Number:
Empirical Formula (Hill Notation):
 C25H22O10
Molecular Weight:
 482.44
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
  Assay
Show More