• USA Home
  • Search Results

  • snglycero3phosphocholine11cadmiumchlorideadduct440546468108911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

sn-Glycero-3-phosphocholine 1:1 cadmium chloride adduct

1 Products
Synonym:   L-α-Glycerophosphorylcholine 1:1 cadmium chloride adduct from egg yolk
Image of <I>sn</I>-<WBR>Glycero-<WBR>3-<WBR>phosphocholine 1:1 cadmium chloride adduct
CAS Number:
Empirical Formula (Hill Notation):
 C8H20NO6P · CdCl2
Molecular Weight:
 440.54
 Beilstein Registry Number:
 4936450
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More