• USA Home
  • Search Results

  • sodium1dodecanesulfonate27238238653011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium 1-dodecanesulfonate

1 Products
Synonym:   1-Dodecanesulfonic acid sodium salt
Image of Sodium 1-dodecanesulfonate
CAS Number:
Linear Formula:
 CH3(CH2)11SO3Na
Molecular Weight:
 272.38
 Beilstein/REAXYS Number:
 3919536
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Product Line
Show More