• USA Home
  • Search Results

  • sodium1naphthalenesulfonate2302213014311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium 1-naphthalenesulfonate

1 Products
Synonym:   1-Naphthalenesulfonic acid sodium salt
Image of Sodium 1-<WBR>naphthalenesulfonate
CAS Number:
Empirical Formula (Hill Notation):
 C10H7NaO3S
Molecular Weight:
 230.22
 Beilstein/REAXYS Number:
 3639038
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More