• USA Home
  • Search Results

  • sodium2oxobutyrate12407201326511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium 2-oxobutyrate

1 Products
Synonym:   2-Oxobutanoic acid sodium salt, 2-Oxobutyric acid sodium salt, Sodium α-ketobutyrate
Image of Sodium 2-oxobutyrate
CAS Number:
Linear Formula:
 CH3CH2COCOONa
Molecular Weight:
 124.07
 Beilstein/REAXYS Number:
 3631701
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More