• USA Home
  • Search Results

  • sodium55diethylbarbiturate2061714402511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium 5,5-diethylbarbiturate

1 Products
Synonym:   5,5-Diethylbarbituric acid sodium salt, Barbitone, Sodium barbital, Veronal sodium salt
Image of Sodium 5,5-<WBR>diethylbarbiturate
CAS Number:
Empirical Formula (Hill Notation):
 C8H11N2NaO3
Molecular Weight:
 206.17
 Beilstein/REAXYS Number:
 3921202
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Purity Grade
  Solubility
Show More