• USA Home
  • Search Results

  • sodiumacetate820312709311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium acetate

20 Products
Synonym:   Acetic acid sodium salt
Image of Sodium acetate
CAS Number:
Linear Formula:
 CH3COONa
Molecular Weight:
 82.03
 Beilstein/REAXYS Number:
 3595639
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss:
 39021906
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Restriction
  Anal. Standard Form
  Special Grade
  UV-Absorption Condition
  Manufacturer designation
  Agency/Method
  Application(s)
  Quality clarifier
  Purity Grade
  Physical Form
  Transition temperature
  Melting Point
  Density
  Autoignition temp
  Product Line
  Suitability
  Loss
  Potency
  UV-Absorption Values
  Solubility
  pH Value
  Assay
Show More

Product #
Description
S2889 Sodium acetate, anhydrous, for molecular biology, ≥99%,
S8750 Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%,
71183 Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT),
241245 Sodium acetate, ACS reagent, ≥99.0%,
1.06268 Sodium acetate, anhydrous for analysis EMSURE® ACS,Reag. Ph Eur,
S1429 Sodium acetate, meets USP testing specifications, anhydrous,
S5636 Sodium acetate, powder, BioReagent, for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%,
32319 Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous,
79714 Sodium acetate, for HPLC, LiChropur, 99.0-101.0% (NT),
S7545 Sodium acetate, BioXtra, ≥99.0%,
791741 Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%,
229873 Sodium acetate, 99.995% trace metals basis,
1.37046 Sodium acetate, anhydrous EMPROVE® EXPERT USP,
1.06264 Sodium acetate, anhydrous 99.99 Suprapur®,
1613407 Sodium acetate, United States Pharmacopeia (USP) Reference Standard,
1.06281 Sodium acetate, anhydrous EMPROVE® ESSENTIAL USP,FCC,E 262,
W302475 Sodium acetate, anhydrous, natural (US), ≥99%, FG, New
908959 Sodium acetate, Redi-Dri, free-flowing, anhydrous, ReagentPlus®, ≥99.0%, New
25023 Sodium acetate, puriss., anhydrous, ≥98%, powder,
PHR2615 Sodium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material, New