• USA Home
  • Search Results

  • sodiumacetatebuffersolution1234512696511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium acetate buffer solution

2 Products
Image of Sodium acetate buffer solution
CAS Number:
 Beilstein/REAXYS Number:
 3786729
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32129211
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Suitability
  Special Grade
  pH Value
  Sterilization
Show More