• USA Home
  • Search Results

  • sodiumacetatetrihydrate13608613190411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium acetate trihydrate

14 Products
Synonym:   Acetic acid sodium salt trihydrate
Image of Sodium acetate trihydrate
CAS Number:
Linear Formula:
 CH3COONa · 3H2O
Molecular Weight:
 136.08
 Beilstein/REAXYS Number:
 3732037
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Application(s)
  Agency/Method
  Restriction
  Suitability
  Purity Grade
  Loss
  Density
  Bulk Density
  Autoignition temp
  Transition temperature
  Melting Point
  Solubility
  Potency
  Product Line
  Special Grade
  Assay
  Physical Form
  pH Value
Show More

Product #
Description
S8625 Sodium acetate trihydrate, ReagentPlus®, ≥99.0%,
236500 Sodium acetate trihydrate, ACS reagent, ≥99%,
1.28205 Sodium acetate trihydrate, EMPROVE® Expert, Ph. Eur., BP, ChP, JP, USP, New
32318 Sodium acetate trihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%,
1.06267 Sodium acetate trihydrate, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
S7670 Sodium acetate trihydrate, BioXtra, ≥99.0%,
S1304 Sodium acetate trihydrate, meets USP testing specifications,
25022 Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al,
71188 Sodium acetate trihydrate, BioUltra, ≥99.5% (NT),
1.06265 Sodium acetate trihydrate, extra pure EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,FCC,E 262,
5.43833 Sodium acetate trihydrate, for HPLC LiChropur,
1613429 Sodium acetate trihydrate, United States Pharmacopeia (USP) Reference Standard,
SX0255 Sodium acetate trihydrate, Meets ACS Specifications GR ACS,
PHR2618 Sodium Acetate Trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material,