• USA Home
  • Search Results

  • sodiumacetylidesuspension4802106626811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium acetylide suspension

1 Products
Synonym:   Ethynyl sodium
Image of Sodium acetylide suspension
CAS Number:
Linear Formula:
 HC≡CNa
Molecular Weight:
 48.02
 Beilstein/REAXYS Number:
 3587232
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More