• USA Home
  • Search Results

  • sodiumarsenatedibasicheptahydrate312011004895011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium arsenate dibasic heptahydrate

2 Products
Synonym:   Disodium hydrogen arsenate heptahydrate, di-Sodium hydrogen arsenate heptahydrate
Image of Sodium arsenate dibasic heptahydrate
CAS Number:
Linear Formula:
 Na2HAsO4 · 7H2O
Molecular Weight:
 312.01
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  pH Value
  Physical Form
Show More