• USA Home
  • Search Results

  • sodiumascorbate1981113403211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium ascorbate

3 Products
Synonym:   (+)-Sodium L-ascorbate, L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt
Image of Sodium ascorbate
CAS Number:
Empirical Formula (Hill Notation):
 C6H7NaO6
Molecular Weight:
 198.11
 Beilstein/REAXYS Number:
 3767246
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More