• USA Home
  • Search Results

  • sodiumazide65012662822811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium azide

6 Products
Synonym:   Hydrazoic acid sodium salt, NSC 3072
Image of Sodium azide
CAS Number:
Linear Formula:
 NaN3
Molecular Weight:
 65.01
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Restriction
  Special Grade
  Vapor pressure
  Loss
  Biological Source
  pH Value
  Product Line
  Melting Point
  Physical Form
  Solubility
Show More