• USA Home
  • Search Results

  • sodiumbenzoate1441053232111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium benzoate

9 Products
Synonym:   Benzoic acid sodium salt
Image of Sodium benzoate
CAS Number:
Linear Formula:
 C6H5COONa
Molecular Weight:
 144.10
 Beilstein/REAXYS Number:
 3572467
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Vapor pressure
  Anal. Standard Form
  pH Value
  Purity Grade
  Application(s)
  Product Line
  Physical Form
  Solubility
  Agency/Method
  Assay
Show More