• USA Home
  • Search Results

  • sodiumbicarbonate840114455811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium bicarbonate

13 Products
Synonym:   Sodium hydrogen carbonate
Image of Sodium bicarbonate
CAS Number:
Linear Formula:
 NaHCO3
Molecular Weight:
 84.01
 Beilstein/REAXYS Number:
 4153970
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Manufacturer designation
  Agency/Method
  Suitability
  Purity Grade
  Application(s)
  Quality clarifier
  pH Value
  Product Line
  Solubility
  Special Grade
  Physical Form
Show More

Product #
Description
S5761 Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture,
S6014 Sodium bicarbonate, ACS reagent, ≥99.7%,
S8875 Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder,
S6297 Sodium bicarbonate, BioXtra, 99.5-100.5%,
13433 Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder,
792519 Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%,
SX0320 Sodium bicarbonate, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
31437 Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%,
S4019 Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%,
71631 Sodium bicarbonate, tested according to Ph. Eur.,
1613655 Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard,
900506 Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%,
NIST2191A Sodium bicarbonate, NIST® SRM® 2191a, pD Standard,