• USA Home
  • Search Results

  • sodiumbisulfatemonohydrate138081003488511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium bisulfate monohydrate

2 Products
Synonym:   Sodium hydrogen sulfate monohydrate
Image of Sodium bisulfate monohydrate
CAS Number:
Linear Formula:
 NaHSO4 · H2O
Molecular Weight:
 138.08
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
  Physical Form
Show More