• USA Home
  • Search Results

  • sodiumborohydride37831694066211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium borohydride

11 Products
Synonym:   Sodium tetrahydridoborate
Image of Sodium borohydride
CAS Number:
Linear Formula:
 NaBH4
Molecular Weight:
 37.83
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Additive
  Purity Grade
  Special Grade
  pH Value
  Vapor pressure
  Transition temperature
  Melting Point
  Explosion limit
  Density
  Composite Dimension
  Bulk Density
  Autoignition temp
  Greener Alternative Principles
  Potency
  Physical Form
Show More