• USA Home
  • Search Results

  • sodiumbromide10289764715611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium bromide

12 Products
Image of Sodium bromide
CAS Number:
Linear Formula:
 NaBr
Molecular Weight:
 102.89
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss:
 38050404
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Values
  Loss
  Density
  Bulk Density
  Vapor pressure
  Quality clarifier
  Agency/Method
  Potency
  Physical Form
  Special Grade
  Solubility
  Product Line
  pH Value
Show More