• USA Home
  • Search Results

  • sodiumbutyrate1100915654711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium butyrate

9 Products
Synonym:   Butyric acid sodium salt
Image of Sodium butyrate
CAS Number:
Linear Formula:
 CH3CH2CH2COONa
Molecular Weight:
 110.09
 Beilstein/REAXYS Number:
 3629439
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Agency/Method
  Suitability
  Solubility
  Special Grade
  Melting Point
  Density
  Biological Source
  Physical Form
  Package quantity
Show More