• USA Home
  • Search Results

  • sodiumcarbonate1059949719811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium carbonate

22 Products
Synonym:   Calcined soda, Carbonic acid disodium salt, Soda ash
Image of Sodium carbonate
CAS Number:
Linear Formula:
 Na2CO3
Molecular Weight:
 105.99
 Beilstein/REAXYS Number:
 4154566
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  pH Value
  Suitability
  pK-Values
  UV-Absorption Values
  Application(s)
  Package quantity
  Manufacturer designation
  Ice Shipping
  Restriction
  Anal. Standard Form
  Purity Grade
  Potency
  Density
  Bulk Density
  Quality clarifier
  Loss
  Solubility
  Product Line
  Agency/Method
  Special Grade
  Assay
  Physical Form
Show More

Product #
Description
S7795 Sodium carbonate, BioXtra, ≥99.0%,
222321 Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules,
223530 Sodium carbonate, powder, ≥99.5%, ACS reagent,
S2127 Sodium carbonate, ReagentPlus®, ≥99.5%,
223484 Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis,
451614 Sodium carbonate, anhydrous, powder, 99.999% trace metals basis,
791768 Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%,
1.06392 Sodium carbonate, anhydrous for analysis EMSURE® ISO,
71363 Sodium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%,
13418 Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance),
71345 Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T),
1.02405 Sodium carbonate, volumetric standard, secondary reference material for acidimetry, traceable to NIST SRM Certipur®,
1.06393 Sodium carbonate, anhydrous, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
1.37014 Sodium carbonate, anhydrous EMPROVE® EXPERT Ph Eur,BP,JP,NF,
1.06395 Sodium carbonate, anhydrous 99.999 Suprapur®,
1.06398 Sodium carbonate, anhydrous EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,
NIST2192A Sodium carbonate, NIST® SRM® 2192a, pD Standard,
SX0395 Sodium carbonate, anhydrous, powder, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
900503 Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%,
900504 Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis,
900505 Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%,
SX0400 Sodium carbonate, anhydrous, granular, Meets ACS Specifications GR ACS,