• USA Home
  • Search Results

  • sodiumcarbonateandydrous1059949719811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium Carbonate Andydrous

1 Products
Synonym:   Calcined soda, Carbonic acid disodium salt, Soda ash, Sodium carbonate
Image of Sodium Carbonate Andydrous
CAS Number:
Linear Formula:
 Na2CO3
Molecular Weight:
 105.99
 Beilstein/REAXYS Number:
 4154566
 EC Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More