• USA Home
  • Search Results

  • sodiumcarbonatedecahydrate28614613202111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium carbonate decahydrate

4 Products
Synonym:   Sal soda, Soda
Image of Sodium carbonate decahydrate
CAS Number:
Linear Formula:
 Na2CO3 · 10H2O
Molecular Weight:
 286.14
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Solubility
  Purity Grade
  Product Line
  Potency
  Melting Point
  Density
  Bulk Density
  Special Grade
  Agency/Method
  Physical Form
  pH Value
Show More