• USA Home
  • Search Results

  • sodiumcarbonatemonohydrate12400596811611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium carbonate monohydrate

3 Products
Image of Sodium carbonate monohydrate
CAS Number:
Linear Formula:
 Na2CO3 · H2O
Molecular Weight:
 124.00
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purity Grade
  Loss
  Special Grade
Show More