• USA Home
  • Search Results

  • sodiumcarbonatesolution1059949719811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium carbonate solution

2 Products
Image of Sodium carbonate solution
CAS Number:
Linear Formula:
 Na2CO3
Molecular Weight:
 105.99
 Beilstein/REAXYS Number:
 4154566
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Application(s)
Show More